Tide Quencher™ 7WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ7WS SE
2111
AATB2111EA
614
EUR
AATB2111
Tide Quencher™ 7WS succinimidyl ester, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 7WS succinimidyl ester
TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
1000,27 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...