Tide Quencher™ 7WS azide, Tide Quencher™ fluoreszierende Probe

Lieferant: AAT BIOQUEST
Achtung

Synonyme: TQ7WS azide

2112
AATB2112EA 614 EUR
AATB2112
Tide Quencher™ 7WS azide, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 7WS azide

TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.


  • Stronger absorption
  • Higher quenching efficiency
  • Versatile reactive forms
  • Excellent oligonucleotide and peptide labeling

Molecular Weight: 984,12 g/mol
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=97dc8787fa501eda182cdd8f884c12eb7721ff38&vid=af0dd4e8b9299c78038acfaeccad1a2ff6eb42f0&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR