Rhod-4™ alkyne
Lieferant: AAT BIOQUEST
21126
AATB21126EA 687 EUR
AATB21126
Rhod-4™ alkyne
Rhod-4™ alkyne
Rhod-4™ alkyne is an excellent building block that can be readily used to prepare a calcium-sensitive bioconjugate for monitoring calcium change spatially for a specific target.
  • Stable for 12 months
Quality tested in-house