Tide Quencher™ 7WS alkyne, Tide Quencher™ fluoreszierende Probe
Lieferant: AAT BIOQUEST
|
Achtung
|
Synonyme:
TQ7WS alkyne
2113
AATB2113EA
614
EUR
AATB2113
Tide Quencher™ 7WS alkyne, Tide Quencher™ fluoreszierende Probe
Tide Quencher™ 7WS alkyne
TQ7WS is designed to be a superior quencher to Cy7, TF7 and Alexa Fluor 750.
- Stronger absorption
- Higher quenching efficiency
- Versatile reactive forms
- Excellent oligonucleotide and peptide labeling
Molecular Weight:
953,3 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...