Cal-520® AM
Lieferant: AAT Bioquest
Achtung
21130 21131
AATB21130EA 307 EUR
AATB21130 AATB21131
Cal-520® AM
Cal-520® AM
Cal-520® AM provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell permeant
Bitte beachten For research use only.
ADR: 5,II