7.1WS acid [TQ7.1WS acid], Tide Quencher™
Lieferant: AAT BIOQUEST
2114
AATB2114EA 410 EUR
AATB2114
7.1WS acid [TQ7.1WS acid], Tide Quencher™
7.1WS acid [TQ7.1WS acid]
Tide Quencher™ 7.1WS (TQ7.1WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy7, Alexa Fluor® 700, Alexa Fluor® 750, iFluor® 700, iFluor® 710, iFluor® 720 and iFluor® 750.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Neuer Artikel bei VWR!
Bestellen
TECHNISCHE DATEN
Environmentally Preferable