Cal-520N™ AM
Lieferant: AAT BIOQUEST
Achtung
21146
AATB21146EA 537 EUR
AATB21146
Cal-520N™ AM
Cal-520N™ AM
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
  • Improved background/signal ratio
  • Suitable for all major fluorescence platforms
  • Cell permeant
Bitte beachten For research use only.
ADR: 5,II