Cal-520N™ AM
Lieferant: AAT BIOQUEST
|
Achtung
|
21146
AATB21146EA
537
EUR
AATB21146
Cal-520N™ AM
Cal-520N™ AM
Cal-520® provides a robust homogeneous fluorescence-based assay tool for detecting intracellular calcium mobilization.
- Improved background/signal ratio
- Suitable for all major fluorescence platforms
- Cell permeant
Bitte beachten For research use only.
ADR:
5,II |