Coelenterazine 400a
Lieferant: AAT BIOQUEST
21167
AATB21167EA
106
EUR
AATB21167
Coelenterazine 400a
Coelenterazine 400a
Coelenterazine 400a is a bisdeoxy derivative of coelenterazine that has an emission of ~395 nm following conversion by Renilla luciferase (Rluc).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only