7.1 CPG [TQ7.1 CPG], Tide Quencher™ 1000 Å
Lieferant: AAT BIOQUEST
2117
AATB2117EA
275
EUR
AATB2117
7.1 CPG [TQ7.1 CPG], Tide Quencher™ 1000 Å
7.1 CPG [TQ7.1 CPG]
Tide Quencher™ 7.1 (TQ7.1) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy7, Alexa Fluor® 700, Alexa Fluor® 750, iFluor® 700, iFluor® 710, iFluor® 720 and iFluor® 750.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only