5-(and-6)-Carboxy SNARF-4F
Lieferant: AAT BIOQUEST
21170
AATB21170EA 673 EUR
AATB21170
5-(and-6)-Carboxy SNARF-4F
5-(and-6)-Carboxy SNARF-4F
SNARF-4F 5-(and-6)-carboxylic acid is a cell-impermeant ratiometric pH indicator with pKa of ∼6.4.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only