5-(and-6)-Carboxy SNARF-4F succinimidyl ester
Lieferant: AAT BIOQUEST
21171
AATB21171EA
538
EUR
AATB21171
5-(and-6)-Carboxy SNARF-4F succinimidyl ester
5-(and-6)-Carboxy SNARF-4F succinimidyl ester
5-(and-6)-Carboxy SNARF-4F Succinimidyl Ester is an excellent amine-reactive building block that can be readily used for developing SNARF-4F pH probes.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only