5-(and-6)-Carboxy SNARF-4F maleimide
Lieferant: AAT BIOQUEST
21172
AATB21172EA 673 EUR
AATB21172
5-(and-6)-Carboxy SNARF-4F maleimide
5-(and-6)-Carboxy SNARF-4F maleimide
5-(and-6)-Carboxy SNARF-4F Maleimide is an excellent thiol-reactive building block that can be readily used for developing SNARF-4F pH probes.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only