Protonex™ Red 670 NHS ester
Lieferant: AAT Bioquest
Protonex™ Red 670 NHS ester is an excellent building block for developing a target-specific conjugate that can be used for monitoring a pH-sensitive biological event.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only