BCFL Acid, RatioWorks™ Superior replacement for BCECF
Lieferant: AAT BIOQUEST
|
Achtung
|
21189
AATB21189EA
152
EUR
AATB21189
BCFL Acid, RatioWorks™ Superior replacement for BCECF
BCFL Acid
Intracellular pH plays an important modulating role in many cellular events, including cell growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, cell adhesion and other cellular processes.
- Improved reliability
- In-house quality testing
- Ratiometric signal
Bitte beachten For research use only.
Molecular Weight:
500 g/mol |
ADR:
5,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...