7.1WS succinimidyl ester [TQ7.1WS SE], Tide Quencher™
Lieferant: AAT BIOQUEST
2119
AATB2119EA
309
EUR
AATB2119
7.1WS succinimidyl ester [TQ7.1WS SE], Tide Quencher™
7.1WS succinimidyl ester [TQ7.1WS SE]
Tide Quencher™ 7.1WS (TQ7.1WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy7, Alexa Fluor® 700, Alexa Fluor® 750, iFluor® 700, iFluor® 710, iFluor® 720 and iFluor® 750.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only