BCECF AM ester, CytoFix™ Optimized for long term cellular pH tracking

Lieferant: AAT BIOQUEST
Achtung

Synonyme: BCECF AM

21200
AATB21200EA 439 EUR
AATB21200
BCECF AM ester, CytoFix™ Optimized for long term cellular pH tracking
BCECF AM ester

Intracellular pH plays an important modulating role in many cellular events, including cell growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, cell adhesion and other cellular processes.


  • Cell permeable
  • Improved cell retention
  • In-house quality testing


Bitte beachten For research use only.

Formel: C₄₂H₄₀O₂₁
Molecular Weight: 808.69 g/mol
MDL: MFCD00036969
CAS-Nummer: 117464-70-7
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR