BCECF AM ester Kalziumindikator

Lieferant: AAT BIOQUEST
Achtung

Synonyme: BCECF AM

21202
AATB21202EA 230 EUR
AATB21202
BCECF AM ester Kalziumindikator
BCECF AM ester

Intracellular pH plays an important modulating role in many cellular events, including cell growth, calcium regulation, enzymatic activity, receptor-mediated signal transduction, ion transport, endocytosis, chemotaxis, cell adhesion and other cellular processes.


  • Excellent intracellular pH measurement

Formel: C₄₂H₄₀O₂₁
Molecular Weight: 808,69 g/mol
MDL: MFCD00036969
CAS-Nummer: 117464-70-7
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acb007751ae0978e1af43e9bb49c651c263c27ab&vid=7c919f1aa07f0e1aaef3a6f9dce62460fa335a8d&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR