Red 600, Protonex™

Lieferant: AAT BIOQUEST
Achtung

21207
AATB21207EA 307 EUR
AATB21207
Red 600, Protonex™
Red 600

Protonex™ Red dye demonstrated pH-dependent fluorescence.


  • pH-dependent fluorescence
  • Multiple applications
  • Suitable for all major fluorescence platforms
  • Enhanced signal at high pH


Bitte beachten For research use only.

Molecular Weight: 698,94 g/mol
ADR: 5,II

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR