Red 600-Latex Bead Konjugat, Protonex™

Lieferant: AAT BIOQUEST
Achtung

21209
AATB21209EA 460 EUR
AATB21209
Red 600-Latex Bead Konjugat, Protonex™
Red 600-Latex Bead

Protonex™ Red-latex bead conjugate demonstrated pH-dependent fluorescence.


  • pH-dependent fluorescence
  • Multiple applications
  • Suitable for all major fluorescence platforms
  • Enhanced signal at high pH


Bitte beachten For research use only.

ADR: 5,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR