PH165 AM, RatioWorks™

Lieferant: AAT BIOQUEST
Achtung

21212
AATB21212EA 292 EUR
AATB21212
PH165 AM, RatioWorks™
PH165 AM

RatioWorks™ PH165 AM is a novel fluorescent probe that can be used to measure intracellular pH in live cells.


  • pH-dependent fluorescence
  • Multiple applications
  • Suitable for all major fluorescence platforms
  • Improved cell retention
  • Ratiometric signal


Bitte beachten For research use only.

Molecular Weight: 583.53 g/mol
ADR: 5 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR