RatioWorks™ PH165 NHS Ester
Lieferant: AAT BIOQUEST
21214
AATB21214EA
292
EUR
AATB21214
RatioWorks™ PH165 NHS Ester
RatioWorks™ PH165 NHS Ester
RatioWorks™ PH165 NHS ester is a novel fluorescent probe that can be used to measure intracellular pH in live cells.
- pH-dependent fluorescence
- Ratiometric detection
- Long-wavelength emission
- Cell impermeable
Bitte beachten For research use only.