RatioWorks™ PH165 NHS Ester
Lieferant: AAT BIOQUEST
21214
AATB21214EA 292 EUR
AATB21214
RatioWorks™ PH165 NHS Ester
RatioWorks™ PH165 NHS Ester
RatioWorks™ PH165 NHS ester is a novel fluorescent probe that can be used to measure intracellular pH in live cells.
  • pH-dependent fluorescence
  • Ratiometric detection
  • Long-wavelength emission
  • Cell impermeable
Bitte beachten For research use only.