Protonex™ Green 500 Dextran
Lieferant: AAT BIOQUEST
21217
AATB21217EA
307
EUR
AATB21217
Protonex™ Green 500 Dextran
Protonex™ Green 500 Dextran
Protonex™ Green dye demonstrated pH-dependent fluorescence.
- pH-dependent fluorescence
- No fluorescence outside the cell
- Excellent for imaging acidic organelles
- Cell impermeable
Bitte beachten For research use only.