Protonex™ Green 500 Dextran
Lieferant: AAT BIOQUEST
21217
AATB21217EA 307 EUR
AATB21217
Protonex™ Green 500 Dextran
Protonex™ Green 500 Dextran
Protonex™ Green dye demonstrated pH-dependent fluorescence.
  • pH-dependent fluorescence
  • No fluorescence outside the cell
  • Excellent for imaging acidic organelles
  • Cell impermeable
Bitte beachten For research use only.