Protonex™ Green 500WS
Lieferant: AAT BIOQUEST
21218
AATB21218EA 307 EUR
AATB21218
Protonex™ Green 500WS
Protonex™ Green 500WS
Protonex™ Green dye demonstrated pH-dependent fluorescence.
  • pH-dependent fluorescence
  • No fluorescence outside the cell
  • Excellent for imaging acidic organelles
  • Cell impermeable
Bitte beachten For research use only.