Protonex™ NIR 770 NHS ester
Lieferant: AAT BIOQUEST
21221
AATB21221EA 538 EUR
AATB21221
Protonex™ NIR 770 NHS ester
Protonex™ NIR 770 NHS ester
Protonex™ NIR 770 NHS ester is the only cell-permeable acidotropic pH dye commercially available for developing NIR pH probes.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only