Protonex™ Red 670-Latex bead Konjugat
Lieferant: AAT BIOQUEST
21222
AATB21222EA
464
EUR
AATB21222
Protonex™ Red 670-Latex bead Konjugat
Protonex™ Red 670-Latex bead
Protonex™ Red 670-Latex Bead Conjugates exhibit unique pH-dependent fluorescence properties.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only