Protonex™ Green 500-Latex bead Konjugat
Lieferant: AAT BIOQUEST
Protonex™ Green 500-Latex Bead Conjugates exhibit distinctive pH-dependent fluorescence characteristics, becoming significantly more fluorescent in acidic conditions, unlike most conventional fluorescent dyes that increase in fluorescence at higher pH levels.
- Stable for 12-months
Quality tested in-house
Bitte beachten For Research Use Only