Protonex™ Green 500-Latex bead Konjugat
Lieferant: AAT BIOQUEST
21223
AATB21223EA 464 EUR
AATB21223
Protonex™ Green 500-Latex bead Konjugat
Protonex™ Green 500-Latex bead
Protonex™ Green 500-Latex Bead Conjugates exhibit distinctive pH-dependent fluorescence characteristics, becoming significantly more fluorescent in acidic conditions, unlike most conventional fluorescent dyes that increase in fluorescence at higher pH levels.
  • Stable for 12-months
Quality tested in-house
Bitte beachten For Research Use Only