Protonex™ Green 500 PEG12 maleimide
Lieferant: AAT BIOQUEST
21224
AATB21224EA 271 EUR
AATB21224
Protonex™ Green 500 PEG12 maleimide
Protonex™ Green 500 PEG12 maleimide
Protonex™ Green 500 PEG12 maleimide is a thiol-reactive, pH-sensitive fluorophore specifically designed for developing pH-responsive bioconjugates to investigate processes such as antibody internalization, endocytosis, and phagocytosis in live cells.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only