5-(and-6)-Carboxy SNARF-1
Lieferant: AAT BIOQUEST
21226
AATB21226EA
470
EUR
AATB21226
5-(and-6)-Carboxy SNARF-1
5-(and-6)-Carboxy SNARF-1
5-(and-6)-Carboxy SNARF-1 is an excellent pH probe for monitoring pH changes in the range of 6.5 and 8.5.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only