5-(and-6)-Carboxy SNARF-1 succinimidyl ester
Lieferant: AAT BIOQUEST
21227
AATB21227EA
538
EUR
AATB21227
5-(and-6)-Carboxy SNARF-1 succinimidyl ester
5-(and-6)-Carboxy SNARF-1 succinimidyl ester
5-(and-6)-Carboxy SNARF-1 Succinimidyl Ester is an excellent amine-reactive building block that can be used for developing SNARF-1 pH probes.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only