5-(and-6)-Carboxy SNARF-1 maleimide
Lieferant: AAT BIOQUEST
21228
AATB21228EA
538
EUR
AATB21228
5-(and-6)-Carboxy SNARF-1 maleimide
5-(and-6)-Carboxy SNARF-1 maleimide
5-(and-6)-Carboxy SNARF-1 Maleimide is an excellent thiol-reactive building block that can be used for developing SNARF-1 pH probes.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only