7.2WS amine [TQ7.2WS amine], Tide Quencher™
Lieferant: AAT BIOQUEST
2123
AATB2123EA 309 EUR
AATB2123
7.2WS amine [TQ7.2WS amine], Tide Quencher™
7.2WS amine [TQ7.2WS amine]
Tide Quencher™ 7.2WS (TQ7.2WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy7, Alexa Fluor® 700, Alexa Fluor® 750, iFluor® 700, iFluor® 710, iFluor® 720 and iFluor® 750.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only