Protonex™ Red 600 PEG12 maleimide
Lieferant: AAT BIOQUEST
Protonex™ Red 600 PEG12 maleimide is a thiol-reactive, pH-sensitive dye that has been specifically engineered for use in developing fluorescent pH probes applicable to live cell assays focusing on processes such as antibody internalization, endocytosis, and phagocytosis.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only