7.2 CPG [TQ7.2 CPG], Tide Quencher™ (500 A)
Lieferant: AAT BIOQUEST
2124
AATB2124EA 275 EUR
AATB2124
7.2 CPG [TQ7.2 CPG], Tide Quencher™ (500 A)
7.2 CPG [TQ7.2 CPG]
Tide Quencher™ 7.2 (TQ7.2) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy7, Alexa Fluor® 700, Alexa Fluor® 750, iFluor® 700, iFluor® 710, iFluor® 720 and iFluor® 750.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only