Protonex™ Red 600 maleimide
Lieferant: AAT BIOQUEST
21240
AATB21240EA
464
EUR
AATB21240
Protonex™ Red 600 maleimide
Protonex™ Red 600 maleimide
Protonex™ Red 600 dye exhibits pH-dependent fluorescence, with enhanced brightness under acidic conditions—opposite to most fluorescent dyes that fluoresce more intensely at higher pH levels.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only