Protonex™ Lyso-Red 670
Lieferant: AAT BIOQUEST
21241
AATB21241EA
410
EUR
AATB21241
Protonex™ Lyso-Red 670
Protonex™ Lyso-Red 670
Lysosomes are membrane-bound organelles containing acid hydrolases that break down biomolecules in an acidic environment (pH 4.5-4.8).
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only