5-(and-6)-Carboxy SNARF-1 azide
Lieferant: AAT BIOQUEST
21243
AATB21243EA
538
EUR
AATB21243
5-(and-6)-Carboxy SNARF-1 azide
5-(and-6)-Carboxy SNARF-1 azide
5-(and-6)-Carboxy SNARF-1 azide is an excellent alkynyl-reactive building block that can be used for developing SNARF-1 pH probes via the well-known click chemistry.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only