5-(and-6)-Carboxy SNARF-1 alkyne
Lieferant: AAT BIOQUEST
5-(and-6)-Carboxy SNARF-1 alkyne is an excellent azido-reactive building block that can be used for developing SNARF-1 pH probes via the well-known click chemistry.
- Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only