5-(and-6)-Carboxy SNARF-5F
Lieferant: AAT BIOQUEST
21245
AATB21245EA 673 EUR
AATB21245
5-(and-6)-Carboxy SNARF-5F
5-(and-6)-Carboxy SNARF-5F
SNARF-5F 5-(and-6)-carboxylic acid is a cell-impermeant ratiometric pH indicator with pKa of ∼7.2.
  • Stable for 24 months
Quality tested in-house
Bitte beachten For Research Use Only