5-(and-6)-Carboxy SNARF-5F succinimidyl ester
Lieferant: AAT BIOQUEST
21246
AATB21246EA
538
EUR
AATB21246
5-(and-6)-Carboxy SNARF-5F succinimidyl ester
5-(and-6)-Carboxy SNARF-5F succinimidyl ester
5-(and-6)-Carboxy SNARF-5F Succinimidyl Ester is an excellent amine-reactive building block that can be readily used for developing SNARF-5F pH probes.
- Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only