5-(and-6)-Carboxy SNARF-5F AM ester
Lieferant: AAT BIOQUEST
21248
AATB21248EA 538 EUR
AATB21248
5-(and-6)-Carboxy SNARF-5F AM ester
5-(and-6)-Carboxy SNARF-5F AM ester
5-(and-6)-Carboxy SNARF-5F AM Ester is an excellent cell-permeable pH probes for ratiometrically monitoring pH changes in live cells.
  • Stable for 12 months
Quality tested in-house
Bitte beachten For Research Use Only