7.2 CPG [TQ7.2 CPG], Tide Quencher™ 1000 Å
Lieferant: AAT BIOQUEST
2125
AATB2125EA
275
EUR
AATB2125
7.2 CPG [TQ7.2 CPG], Tide Quencher™ 1000 Å
7.2 CPG [TQ7.2 CPG]
Tide Quencher™ 7.2 (TQ7.2) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy7, Alexa Fluor® 700, Alexa Fluor® 750, iFluor® 700, iFluor® 710, iFluor® 720 and iFluor® 750.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)