6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ) Chlorid-Indikator

Lieferant: AAT BIOQUEST
Achtung

Synonyme: SPQ, 3-(6-Methoxyquinolin-1-ium-1-yl)propane-1-sulphonate

21252
AATB21252EA 120 EUR
AATB21252
6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ) Chlorid-Indikator
6-Methoxy-N-(3-sulphopropyl)quinolinium (SPQ)

A chloride indicator that has the fluorescence dependent of chloride concentration.


  • Chloride-dependent fluorescence
  • Well-cited

Formel: C₁₅H₁₅NO₄S
Molecular Weight: 281 g/mol
MDL: MFCD00037592
CAS-Nummer: 83907-40-8
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1cad79905ff5a68106d8dbd437e98f4ea5886cbc&vid=0ec609642866effb53e550d2ceef5231d1125b67&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR