Zinquin ethyl ester, Ultrareine Qualität Zinkindikator

Lieferant: AAT BIOQUEST
Achtung

Synonyme: 2-[[2-Methyl-8-[[(4-methylphenyl)sulphonyl]amino]-6-quinolinyl]oxy]-acetic acid ethyl ester

21253
AATB21253EA 152 EUR
AATB21253
Zinquin ethyl ester, Ultrareine Qualität Zinkindikator
Zinquin ethyl ester

Zinquin ethyl ester is a lipophilic, zinc-sensitive, cell-permeable fluorescent probe.


  • Cell permeable probe
  • Ultrapure grade

Formel: C₂₁H₂₂N₂O₅S
Molecular Weight: 414,47 g/mol
CAS-Nummer: 181530-09-6
ADR: 5,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da60aa89f066595a66db0f1ec8d20c363d8cdc17&vid=adf08cbd69dbf455bd377a67e292957231e32063&prid=web&clid=SAPDAM

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR