MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) Chlorid-Indikator

Lieferant: AAT BIOQUEST
Achtung

Synonyme: (6-Methoxyquinolinio)acetic acid ethyl ester bromide, 1-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide, 1-(2-Ethoxy-2-oxoethyl)-6-methoxyquinolin-1-ium bromide, MQAE

21255
AATB21255EA 120 EUR
AATB21255
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide) Chlorid-Indikator
MQAE (N-(Ethoxycarbonylmethyl)-6-methoxyquinolinium bromide)

A chloride indicator that has the fluorescence dependent of chloride concentration.


  • Chloride-dependent fluorescence
  • Well-cited

Formel: C₁₄H₁₆NO₃Br
Molecular Weight: 326.19 g/mol
MDL: MFCD00467854
CAS-Nummer: 162558-52-3
ADR: 5 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0f5305cc3de85361d4c4c95acde8e4ec166232fe&vid=ae13346da4c5179f745e3b48e5270864728fb000&prid=web&clid=SAPDAM

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR