Zinquin AM, Ultrareine Qualität
Lieferant: AAT Bioquest
21261
AATB21261EA 307 EUR
AATB21261
Zinquin AM, Ultrareine Qualität
Zinquin AM
Zinquin AM ester is a lipophilic, zinc-sensitive, cell-permeable fluorescent probe.
  • Cell permeable
  • Zinc-dependent fluorescence
  • Multiple applications
Bitte beachten For research use only.