Metal Fluor™ Zn-520 potassium salt
Lieferant: AAT Bioquest
21262
AATB21262EA 307 EUR
AATB21262
Metal Fluor™ Zn-520 potassium salt
Metal Fluor™ Zn-520 potassium salt
Metal Fluor™ Zn-520 is designed for detection of higher zinc ion concentrations that are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists (0.05 to 50 µM) with minimal interfering calcium sensitivity.
  • Cell impermeable
  • Zinc-dependent fluorescence
  • Multiple applications
Bitte beachten For research use only.