Metal Fluor™ Zn-520 AM
Lieferant: AAT BIOQUEST
21263
AATB21263EA 307 EUR
AATB21263
Metal Fluor™ Zn-520 AM
Metal Fluor™ Zn-520 AM
Metal Fluor™ Zn-520 is designed for detection of higher zinc ion concentrations that are present in synaptic vesicles and released in response to electrical stimulation or excitotoxic agonists (0.05 to 50 µM) with minimal interfering calcium sensitivity.
  • Cell permeable
  • Zinc-dependent fluorescence
  • Multiple applications
Bitte beachten For research use only.