7.2WS azide [TQ7.2WS azide], Tide Quencher™
Lieferant: AAT BIOQUEST
2128
AATB2128EA 309 EUR
AATB2128
7.2WS azide [TQ7.2WS azide], Tide Quencher™
7.2WS azide [TQ7.2WS azide]
Tide Quencher™ 7.2WS (TQ7.2WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of common NIR fluorophores such as Cy7, Alexa Fluor® 700, Alexa Fluor® 750, iFluor® 700, iFluor® 710, iFluor® 720 and iFluor® 750.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only