8WS acid [TQ8WS acid], Tide Quencher™
Lieferant: AAT BIOQUEST
2130
AATB2130EA 410 EUR
AATB2130
8WS acid [TQ8WS acid], Tide Quencher™
8WS acid [TQ8WS acid]
Tide Quencher™ 8WS (TQ8WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of IR fluorophores such as ICG, iFluor® 820, iFluor® 840 and iFluor® 860.
  • Stable for 12 months
  • Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only