8WS acid [TQ8WS acid], Tide Quencher™
Lieferant: AAT BIOQUEST
2130
AATB2130EA
410
EUR
AATB2130
8WS acid [TQ8WS acid], Tide Quencher™
8WS acid [TQ8WS acid]
Tide Quencher™ 8WS (TQ8WS) is a non-fluorescent molecule designed to efficiently quench the fluorescence of IR fluorophores such as ICG, iFluor® 820, iFluor® 840 and iFluor® 860.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only