8 CPG [TQ8 CPG], Tide Quencher™ (500 A)
Lieferant: AAT BIOQUEST
2132
AATB2132EA
275
EUR
AATB2132
8 CPG [TQ8 CPG], Tide Quencher™ (500 A)
8 CPG [TQ8 CPG]
Tide Quencher™ 8 (TQ8) is a non-fluorescent molecule designed to efficiently quench the fluorescence of IR fluorophores such as ICG, iFluor® 820, iFluor® 840 and iFluor® 860.
- Stable for 12 months
- Optimized for FRET applications
Quality tested in-house
Bitte beachten For Research Use Only